Learn More
Volatile GC/MS Calibration Standard (BFB) (For EPA 524.2, 624, 8240, 8260, CLP)
Catalog No. USSTS110N1
Encompass
Change view
Click to view available options
Quantity:
1 x 1 mL
Catalog No. Quantity
US-STS-110N-1 1 x 1 mL
1 options
Catalog No. US-STS-110N-1 Supplier Agilent Technologies Supplier No. STS110N1
4-Bromofluorobenzene at 2000μg/mL in methanol
Specifications
Chemical Name or Material Volatile GC/MS Calibration Standard (BFB)
Packaging Ampule
Methods For EPA Methods 524.2, 624, 8240
Quantity 1 x 1 mL