Learn More
AccuStandard, Inc Volatile Organic Compounds-Liquids, 1 mL
SDP
Catalog No. NC1653849
Encompass
Matrix: 2000 ug/mL in Methanol. Number of Components: 55.
Catalog No. NC1653849 Supplier AccuStandard, Inc Supplier No. M502AR310X
Matrix: 2000 ug/mL in Methanol. Number of Components: 55.