Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Volatile Organic Compounds 1, Proficiency Testing Sample, MilliporeSigma™ Supelco™

Catalog No. 111022987 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
1.2 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No.
11-102-2987 1.2 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 11-102-2987 Supplier MilliporeSigma Supelco Supplier No. PE12501.2ML

missing translation for 'validMainframeMsg'

This proficiency testing sample is produced in accordance with ISO/IEC 17043:2010.

TRUSTED_SUSTAINABILITY

Specifications

Flash Point 11°C (Closed Cup, Solvent)
Packaging Ampule
Quantity 1.2 mL
UN Number UN1230 - Class 3 - PG 2 - Methanol, solution
Grade Proficiency Testing Sample
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.