Learn More
Perkin Elmer US LLC Volatile Sample Pan Kit - Tin, Qty. 400
Supplier:  Perkin Elmer US LLC N2410320
Encompass_Preferred
This pan is ideal for holding volatile liquid or solid samples, including viscous materials. This kit requires, but does not include, the Volatile Sample Sealer (02190061).
Catalog No. 50-211-3153