Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Volatiles Mix 1 (VOA-1) (For EPA 601, 624)

Catalog No. USEPA2141N1
Encompass
Change view
Click to view available options
Quantity:
1 x 1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
US-EPA2141N1 1 x 1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. US-EPA2141N1 Supplier Agilent Technologies Supplier No. EPA2141N1
May include imposed supplier surcharges.
Add to Cart
Add to Cart

Specifications

Chemical Name or Material Volatiles Mix 1 (VOA-1)
Packaging Ampule
Quantity 1 x 1 mL
Concentration or Composition (by Analyte or Components) Each component at 50μg/mL in Methanol:
Carbon Tetrachloride
Chlorobenzene
1,3-Dichlorobenzene
1,4-Dichlorobenzene
1,2-Dichloroethane
1,1-Dichloroethene
trans-1,2-Dichloroethane
1,2-Dichloropropane
Ethylbenzene
Tetrachloroethene
1,1,2-Trichloroethane
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.