Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3B Scientific™ Volcanic Rocks and Minerals Collection
SDPSCI_ED

Catalog No. S20724 Shop All 3B Scientific Products
Change view
Click to view available options
Quantity:
1
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
S20724 1
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. S20724 Supplier 3B Scientific™ Supplier No. U72010
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

3B Scientific™ Volcanic Rocks and Minerals Collection contains volcanic rocks and minerals. Includes case and key card.

  • Volcanic rocks: basalt, phonolite, pitchstone, rhyolite
  • Lava: Lava from Vesuvius, basaltic lava and rhyoltic lava
  • Pyroclasts: lapilli, volcanic ash, pumice stone
  • Minerals: anorthite, anorthoclase, augite, cristobalite, hauyne, leucite, natrolite, nepheline, pickeringite, sanidine, sulphur, thaumasite, tridymite, obsidian

Specifications

Includes 24 Rocks
Width (Metric) 23 cm
Height (Metric) 6 cm
Quantity 1
Material Plastic, Rock
Color Blue, Gray
Languages English
Subject Earth Science
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.