Learn More
3B Scientific™ Volcanic Rocks, Set of 3
SDPSCI_ED
Catalog No. S20723 Shop All 3B Scientific Products
Change view
Click to view available options
Quantity:
1
Catalog No. Quantity
S20723 1
1 options
Catalog No. S20723 Supplier 3B Scientific™ Supplier No. U72035
Add to Cart
Add to Cart
3B Scientific™ Volcanic Rocks Set consists of three bags, each containing ten pieces of either lava rock, obsidian or pumice stone
Specifications
Includes 30 Rocks, 3 Bags
Width (Metric) 5 cm
Height (Metric) 15 cm
Quantity 1
Material Cloth, Rock
Color White, Black
Languages English
Subject Earth Science