Learn More
Medchemexpress LLC Volinanserin-d4 hydrochloride | 1217617-73-6 | 98.6% | 413.95 | 1 MG
SDP
Supplier:  Medchemexpress LLC HY14940S1MG
Encompass_Preferred
Volinanserin-d4 (hydrochloride) is the deuterium labeled Volinanserin hydrochloride. Volinanserin is a potent and selective antagonist of the 5-HT2 receptor, with a Ki of 0.36 nM. It exhibits 300-fold selectivity for the 5-HT2 receptor over 5-HT1c, alpha-1, and DA D2 receptors. Volinanserin also demonstrates antipsychotic activity.
  • Can be utilized as a tracer.
  • Can serve as an internal standard for quantitative analysis using techniques such as NMR, GC-MS, or LC-MS.
  • Stable heavy isotopes of hydrogen, carbon, and other elements, like deuterium, are incorporated into drug molecules for quantitation during drug development.
Catalog No. 50-003-59541