Learn More
Cayman Chemical VolInserIn 10mg
SDP
Catalog No. 502969597
Encompass_Preferred
A potent and selective antagonist of the serotonin receptor 5-HT2A (Ki = 0.36-0.85 nM); weakly binds 5-HT2C, α1-adrenergic, and sigma receptors (Ki = 88, 128, and 87 nM) and much less effectively associates with other serotonin, dopamine, adrenergic, muscarinic acetylcholine receptors and monoamine transporters
Catalog No. 50-296-9597 Supplier Cayman Chemical Supplier No. 1593610 MG
A potent and selective antagonist of the serotonin receptor 5-HT2A (Ki = 0.36-0.85 nM); weakly binds 5-HT2C, α1-adrenergic, and sigma receptors (Ki = 88, 128, and 87 nM) and much less effectively associates with other serotonin, dopamine, adrenergic, muscarinic acetylcholine receptors and monoamine transporters