Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

BINDER™ Voltage Converter
SDP

Catalog No. 08111188 Shop All BINDER Inc. Products
Change view
Click to view available options
Material:
Steel
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Material
08-111-188 Steel
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 08-111-188 Supplier BINDER™ Supplier No. 80090821

Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.

For Use With: KBF 115, 240, 720, 1020 Constant Climate Chambers

  • To operate the device at a nominal voltage of 120V, 60 Hz
  • Provided individually

Specifications

Description Converter, voltage - 240 V option model, to operate the device (at a nominal voltage of 120 V 1 ph 60 Hz)
For Use With (Equipment) BINDER Chambers Series KBF. KBF P, KBF LQC
Material Steel
Type Voltage Converter
Voltage 120 V
Frequency 60 Hz
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.