Learn More
XRF Scientific Americas Inc. Volatile Matter Crucible 25ml, 36 x 35mm
Supplier:  XRF Scientific Americas Inc. GVM25+LID
Encompass
36X35mm crucible with a 25 ml capacity with volatile lid. Standard lid with no ventilation hole.These 99.95% pure platinum crucibles are available in tall forms, and used for volatile matter analysis. Due to their high temperature strenght, ductility, corrosion and oxidation, excellent resistance to chemical attacks by acids and fusion mixtures resistance; these platinum crucibles are critical when high temperatures & resistance to chemical attacks are required.
Catalog No. NC1231732