Learn More
Volu Sol Hematology Buffer (pH 6.8), 5 gal / 20 L, Volu-Sol™
Supplier: Volu Sol VWB640
Phosphate Buffer, intended for use as a buffer in various Romanowsky type staining procedures (Wright, Wright-Giemsa, Giemsa, May Grunwald, Jenner, and Leishman)