Learn More
Volu Sol Volu-Sol Definitive DCM Slide Stainer
SDP
Supplier:  Volu Sol DCM100
Encompass_Preferred
Automated benchtop slide stainer. The Definitive DCM is variable slide stainer allowing you to stain 1 up to 4 slides at once for consistent, reliable results. Able to provide premium results in stain times as quick as 1 minute. Excellent for quick hematology and cytology.
Catalog No. NC2015960
Add to Cart
Request a Quote