Learn More
Vee Gee Volumetric Flask, Class A, Glass Stopper, 200 mL
Catalog No. 502250381
Encompass_Preferred
The SIBATA Class A Volumetric Flask with ground glass stopper features machine-blown body and heavy-beaded, heavy-tubing neck that is tooled for standard taper glass stopper.• Designed """"To Contain"""" with a temperature of calibration at 20°C• Constructed of borosilicate glass with a low coefficient of expansion• Graduation line is sharply defined and of uniform width• Flask has a large marking square and includes a glass stopper• Meets ASTM E-288 Class A Specifications & USP Standards for Volumetric Flasks
This item is not returnable. View return policy
Catalog No. 50-225-0381 Supplier Vee Gee Supplier No. 2306A200
The SIBATA Class A Volumetric Flask with ground glass stopper features machine-blown body and heavy-beaded, heavy-tubing neck that is tooled for standard taper glass stopper.• Designed """"To Contain"""" with a temperature of calibration at 20°C• Constructed of borosilicate glass with a low coefficient of expansion• Graduation line is sharply defined and of uniform width• Flask has a large marking square and includes a glass stopper• Meets ASTM E-288 Class A Specifications & USP Standards for Volumetric Flasks