Learn More
Bellco Glass, Inc. Volumetric Flask,1000mL To Contain
SDP
Catalog No. 501945496
Encompass_Preferred
Manufactured from Type 1 Class A Borosilicate Glass. Ideal for use to prepare solutions, and to contain. Flat bottom bulb with a elongated neck with permanent graduation line and marking area. Volumetric flasks are calibrated for a particular temperature usually 20 degree Celsius.Specifications:Type 1 Class A Borosilicate GlassSize: 10mlStopper Size: 22Dimensions: 122x327mmCase Quantity: 2
Catalog No. 50-194-5496 Supplier Bellco Glass, Inc. Supplier No. 252501000
Manufactured from Type 1 Class A Borosilicate Glass. Ideal for use to prepare solutions, and to contain. Flat bottom bulb with a elongated neck with permanent graduation line and marking area. Volumetric flasks are calibrated for a particular temperature usually 20 degree Celsius.Specifications:Type 1 Class A Borosilicate GlassSize: 10mlStopper Size: 22Dimensions: 122x327mmCase Quantity: 2