Learn More
Cole-Parmer Cole-Parmer elements Volumetric Flask, Glass, with Glass Stopper, 1000 mL; 1/PK
Catalog No. NC1457180 Shop All Cole-Parmer Products
Encompass_Preferred
These Cole-Parmer elements™ volumetric flasks are constructed of quality borosilicate 3.3 glass. These flasks are calibrated "To Contain" (IN) by DIN standard at 20C. Flasks can withstand a wide temperature range from 50 to 550C.
Catalog No. NC1457180 Supplier Cole-Parmer Supplier No. 3450287
These Cole-Parmer elements™ volumetric flasks are constructed of quality borosilicate 3.3 glass. These flasks are calibrated "To Contain" (IN) by DIN standard at 20C. Flasks can withstand a wide temperature range from 50 to 550C.