Learn More
Cole-Parmer Cole-Parmer elements Volumetric Flask, Glass, with Glass Stopper, 100 mL; 2/PK
Catalog No. NC1457173 Shop All Cole-Parmer Products
Encompass
These Cole-Parmer elements™ volumetric flasks are constructed of quality borosilicate 3.3 glass. These flasks are calibrated "To Contain" (IN) by DIN standard at 20°C. Flasks can withstand a wide temperature range from –50 to 550°C.
Catalog No. NC1457173 Supplier Cole-Parmer Supplier No. 3450283
These Cole-Parmer elements™ volumetric flasks are constructed of quality borosilicate 3.3 glass. These flasks are calibrated "To Contain" (IN) by DIN standard at 20°C. Flasks can withstand a wide temperature range from –50 to 550°C.