Learn More
Bellco Glass, Inc. Volumetric Flask,100mL To Contain
SDP
Catalog No. 501945492
Encompass_Preferred
Manufactured from Type 1 Class A Borosilicate Glass. Ideal for use to prepare solutions, and to contain. Flat bottom bulb with a elongated neck with permanent graduation line and marking area. Volumetric flasks are calibrated for a particular temperature usually 20 degree Celsius.Specifications:Type 1 Class A Borosilicate GlassSize: 10mlStopper Size: 13Dimensions: 58x181mmCase Quantity: 4
Catalog No. 50-194-5492 Supplier Bellco Glass, Inc. Supplier No. 252500100
Manufactured from Type 1 Class A Borosilicate Glass. Ideal for use to prepare solutions, and to contain. Flat bottom bulb with a elongated neck with permanent graduation line and marking area. Volumetric flasks are calibrated for a particular temperature usually 20 degree Celsius.Specifications:Type 1 Class A Borosilicate GlassSize: 10mlStopper Size: 13Dimensions: 58x181mmCase Quantity: 4