Learn More
Foxx Life Sciences Borosil® Volumetric Flask Class A with Interchangeable PP Stopper (14/23), Batch Certificate, 250 mL, 10/CS
Catalog No. 501720271
Encompass_Preferred
Borosil® Class A Volumetric Flasks are individually calibrated by automatic, laser-controlled calibration equipment. Supplied with standard PP stoppers. Flasks comply with ASTM E-288-ClassA. 20mL option is not covered in ASTM. 5mL, 10mL, and 20mL options have a trapezoidal shape for improved stability. Available from 5mL to 2000mL. In collaboration with Borosil Glass Works, Foxx Life Sciences is proud to bring the outstanding quality of Borosil® Type 3.3 Borosilicate glassware to North America for the first time. Manufactured from the same USP Type 1 borosilicate glass that North American scientists have come to trust, Borosil® Laboratory Glassware offers unparalleled mechanical strength, clarity, and reliability at competitive prices. And thanks to a unique annealing process, Borosil® Glassware is stress-free and incredibly resistant to thermal shock.
Catalog No. 50-172-0271 Supplier Foxx Life Sciences Supplier No. 5645021A
Borosil® Class A Volumetric Flasks are individually calibrated by automatic, laser-controlled calibration equipment. Supplied with standard PP stoppers. Flasks comply with ASTM E-288-ClassA. 20mL option is not covered in ASTM. 5mL, 10mL, and 20mL options have a trapezoidal shape for improved stability. Available from 5mL to 2000mL. In collaboration with Borosil Glass Works, Foxx Life Sciences is proud to bring the outstanding quality of Borosil® Type 3.3 Borosilicate glassware to North America for the first time. Manufactured from the same USP Type 1 borosilicate glass that North American scientists have come to trust, Borosil® Laboratory Glassware offers unparalleled mechanical strength, clarity, and reliability at competitive prices. And thanks to a unique annealing process, Borosil® Glassware is stress-free and incredibly resistant to thermal shock.