Learn More
Bellco Glass, Inc. Volumetric Flask,250mL To Contain
SDP
Catalog No. 501945494
Encompass_Preferred
Manufactured from Type 1 Class A Borosilicate Glass. Ideal for use to prepare solutions, and to contain. Flat bottom bulb with a elongated neck with permanent graduation line and marking area. Volumetric flasks are calibrated for a particular temperature usually 20 degree Celsius.Specifications:Type 1 Class A Borosilicate GlassSize: 10mlStopper Size: 16Dimensions: 78x239mmCase Quantity: 4
Catalog No. 50-194-5494 Supplier Bellco Glass, Inc. Supplier No. 252500250
Manufactured from Type 1 Class A Borosilicate Glass. Ideal for use to prepare solutions, and to contain. Flat bottom bulb with a elongated neck with permanent graduation line and marking area. Volumetric flasks are calibrated for a particular temperature usually 20 degree Celsius.Specifications:Type 1 Class A Borosilicate GlassSize: 10mlStopper Size: 16Dimensions: 78x239mmCase Quantity: 4