Learn More
Cole-Parmer Essentials Volumetric Flask, Glass, with Glass Stopper, 25 mL; 2/PK
Catalog No. NC2014844 Shop All Cole-Parmer Products
Encompass_Preferred
These Cole-Parmer Essentials volumetric flasks are constructed of quality borosilicate 3.3 glass. These flasks are calibrated "To Contain" (IN) by DIN standard at 20C. Flasks can withstand a wide temperature range from 50 to 550C.
Catalog No. NC2014844 Supplier Cole-Parmer Supplier No. 3450281
These Cole-Parmer Essentials volumetric flasks are constructed of quality borosilicate 3.3 glass. These flasks are calibrated "To Contain" (IN) by DIN standard at 20C. Flasks can withstand a wide temperature range from 50 to 550C.