Learn More
Chemglass Life Sciences Volumetric Flask, 750mL, Graduated 680 to 780mL, Funnel Pour Top, Wine, Serialized
SDP
Catalog No. 501531202
Encompass_Preferred
Volumetric Flask, 750mL, Graduated 680 to 780mL, Funnel pour Top, Wine, Serialized. Manufactured from borosilicate glass these wine testing volumetric flasks have wide bases and a flat bottoms for stability. Flask neck is graduated in 1mL subdivisions from 680 to 780mL. The long neck is calibrated To Contain at 20C and markings indicate the volume of liquid when filled to that graduation line. Scale is blue, serial number is black. Funnel top includes a pouring spout. Tested and calibrated to provide precise measurements for checking fill volumes, this volumetric flask is certified for wine bottling according to TTB standards and is also very useful in the wine testing step of the wine making process.
Catalog No. 50-153-1202 Supplier Chemglass Life Sciences Supplier No. CG1628750
Volumetric Flask, 750mL, Graduated 680 to 780mL, Funnel pour Top, Wine, Serialized. Manufactured from borosilicate glass these wine testing volumetric flasks have wide bases and a flat bottoms for stability. Flask neck is graduated in 1mL subdivisions from 680 to 780mL. The long neck is calibrated To Contain at 20C and markings indicate the volume of liquid when filled to that graduation line. Scale is blue, serial number is black. Funnel top includes a pouring spout. Tested and calibrated to provide precise measurements for checking fill volumes, this volumetric flask is certified for wine bottling according to TTB standards and is also very useful in the wine testing step of the wine making process.