Learn More
CHEMSCIENCE Volumetric Flask, 75mL, Serialized and Certified, Heavy Duty, Wide Mouth, PE Stopper- 16, Class A
Catalog No. NC2117270
Encompass_Preferred
Volumetric Flask, 75mL, Serialized and Certified, Heavy Duty, Wide Mouth, PE Stopper- 16, Class A
This item is not returnable. View return policy
Catalog No. NC2117270 Supplier CHEMSCIENCE Supplier No. 129.503.04A
Volumetric Flask, 75mL, Serialized and Certified, Heavy Duty, Wide Mouth, PE Stopper- 16, Class A