Learn More
Chemglass Life Sciences 25mL Volumetric Flask, Class A, #9 Outer Stopper Neck, Mixing Bulb
SDP
Catalog No. 501531186
Encompass_Preferred
25mL Volumetric Flask, Class A, #9 Outer Stopper Neck, Mixing Bulb. Similar to CG-1600 but with the addition of a mixing bulb between the calibration line and the stopper. The mixing bulb contains approximately one tenth of the flask capacity and aids in the mixing or dissolving of the flask contents. Supplied complete with one solid glass pennyhead stopper. For additional stoppers, see CG-3018.
Catalog No. 50-153-1186 Supplier Chemglass Life Sciences Supplier No. CG160102
25mL Volumetric Flask, Class A, #9 Outer Stopper Neck, Mixing Bulb. Similar to CG-1600 but with the addition of a mixing bulb between the calibration line and the stopper. The mixing bulb contains approximately one tenth of the flask capacity and aids in the mixing or dissolving of the flask contents. Supplied complete with one solid glass pennyhead stopper. For additional stoppers, see CG-3018.