Learn More
Vee Gee The SIBATA reusable and autoclavable volumetric glass pipets are meticulously manufactured for tolerances and accuracy. The glass is a superior material for measuring samples with a wide variety of chemical compounds.
Supplier:  Vee Gee 2040A5
Encompass_Preferred
• Meet ASTM E-969 Class A Specifications and USP Standards for volumetric glassware • Capacity: 5.0 mL • Tolerance: ±0.01 mL • Color Code: White • Approx. Length: 375 mm • Autoclavable
Catalog No. 50-225-0360