Learn More
Vee Gee The SIBATA reusable and autoclavable volumetric glass pipets are meticulously manufactured for tolerances and accuracy. The glass is a superior material for measuring samples with a wide variety of chemical compounds.
Catalog No. 502250361
Encompass_Preferred
• Meet ASTM E-969 Class A Specifications and USP Standards for volumetric glassware • Capacity: 50.0 mL • Tolerance: ±0.05 mL • Color Code: Red • Approx. Length: 540 mm • Autoclavable
This item is not returnable. View return policy
Catalog No. 50-225-0361 Supplier Vee Gee Supplier No. 2040A50
• Meet ASTM E-969 Class A Specifications and USP Standards for volumetric glassware • Capacity: 50.0 mL • Tolerance: ±0.05 mL • Color Code: Red • Approx. Length: 540 mm • Autoclavable