Learn More
Foxx Life Sciences Borosil Reusable Class A Volumetric Pipettes (Pipets), 2mL, 10/CS
Catalog No. 501542890
Encompass_Preferred
Borosil Reusable Class A Volumetric Pipettes are graduated using a resilient white enamel & calibrated for delivery (TD). Each pipette is individually calibrated by automatic, laser-controlled calibration equipment. Manufactured with sturdy, consistent wall thickness. Reusable. Uniform inner bore diameter for smooth flow. Beveled tips tips resist chipping. Pipettes can be autoclaved and, if necessary, flame-sterilized. Borosil Volumetric Pipettes are graduated for delivery from the capacity mark to the tip, without "blowing out" the droplet remaining in the tip after dispensing. Borosil Class A Volumetric Pipettes comply with ASTM E-969-02. Avail. in sizes ranging from 1mL to 100mL. 1.5mL and 2.5mL options are not covered by ASTM standards. Manufactured from the same USP Type 1 borosilicate glass that North American scientists have come to trust, Borosil Laboratory Glassware offers unparalleled mechanical strength, clarity & reliability at competitive prices.
Catalog No. 50-154-2890 Supplier Foxx Life Sciences Supplier No. 7101002A
Borosil Reusable Class A Volumetric Pipettes are graduated using a resilient white enamel & calibrated for delivery (TD). Each pipette is individually calibrated by automatic, laser-controlled calibration equipment. Manufactured with sturdy, consistent wall thickness. Reusable. Uniform inner bore diameter for smooth flow. Beveled tips tips resist chipping. Pipettes can be autoclaved and, if necessary, flame-sterilized. Borosil Volumetric Pipettes are graduated for delivery from the capacity mark to the tip, without "blowing out" the droplet remaining in the tip after dispensing. Borosil Class A Volumetric Pipettes comply with ASTM E-969-02. Avail. in sizes ranging from 1mL to 100mL. 1.5mL and 2.5mL options are not covered by ASTM standards. Manufactured from the same USP Type 1 borosilicate glass that North American scientists have come to trust, Borosil Laboratory Glassware offers unparalleled mechanical strength, clarity & reliability at competitive prices.