Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Von Hippel-Lindau protein (human) | >90.0% | 10 UG
SDP

Catalog No. 5000292265
Encompass_Preferred

Recombinant full-length human von Hippel-Lindau (VHL) protein with an N-terminal 6xHis tag, supplied as a lyophilized powder for research applications. The protein corresponds to residues M1-D213 (Accession P40337), is expressed in E. coli, and is provided with low endotoxin and high purity suitable for biochemical and binding assays.

  • Source organism: human.
  • Sequence length: full length (M1-D213).
  • Tag: N-terminal 6xHis for affinity purification.
  • Expression system: E. coli.
  • Purity: >90.0% by reducing SDS-PAGE.
  • Molecular weight: ~34 kDa.
  • Endotoxin: <1 EU/μg.
  • Appearance and formulation: lyophilized powder with trehalose-containing buffer.
  • Storage and reconstitution: store lyophilized at -20°C; do not reconstitute below 100 μg/mL in ddH2O; freeze aliquots for long-term storage.

Catalog No. 50-002-92265 Supplier Medchemexpress LLC Supplier No. HYP7612410UG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Recombinant full-length human von Hippel-Lindau (VHL) protein with an N-terminal 6xHis tag, supplied as a lyophilized powder for research applications. The protein corresponds to residues M1-D213 (Accession P40337), is expressed in E. coli, and is provided with low endotoxin and high purity suitable for biochemical and binding assays.

  • Source organism: human.
  • Sequence length: full length (M1-D213).
  • Tag: N-terminal 6xHis for affinity purification.
  • Expression system: E. coli.
  • Purity: >90.0% by reducing SDS-PAGE.
  • Molecular weight: ~34 kDa.
  • Endotoxin: <1 EU/μg.
  • Appearance and formulation: lyophilized powder with trehalose-containing buffer.
  • Storage and reconstitution: store lyophilized at -20°C; do not reconstitute below 100 μg/mL in ddH2O; freeze aliquots for long-term storage.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.