Learn More
StatLab Von Kossa MasterTech Special Stain Kit
SDP
Catalog No. 22000054
Change view
Click to view available options
100 mL
Catalog No.
22-000-054 100 mL
1 options
This item is not returnable. View return policy
Harmful if swallowed, in contact with skin, or if inhaled
Catalog No. 22-000-054 Supplier StatLab Supplier No. KTVKO
Add to Cart
Edge
Add to Cart
This item is not returnable. View return policy
Harmful if swallowed, in contact with skin, or if inhaled
Stains for calcium deposits in just 20 minutes
  • Silver Nitrate serves as the primary stain, reacting with the anions and ultra-violet light to produce brown to black calcium salts against pink to red cytoplasm and nuclei, counterstained by Nuclear Fast Red
  • 100 mL Stain Kit yields between 20 and 32 slides
  • A MasterTech Special Stain Kit
Specifications
Product Type Stain Kit