Learn More
Polysciences, Inc. Von Kossa Method for Calcium Kit
SDP
Catalog No. NC9934501
Encompass_Preferred
Von Kossa Method for Calcium Kit demonstrates calcium phosphate and calcium carbonate salts. Von Kossa Method for Calcium Kit can reveal calcium deposits seen in metabolic diseases, Pagets disease, renal osteodystrophy, hyperparathyroidism, infarction (Gandy Gamna bodies), necrotic areas associated with TB, atheroma in blood vessels and Malakoplakia in the bladder (Michaelis-Gutman bodies). It is also used as a bone stain to indicate Osteomalacia.
Catalog No. NC9934501 Supplier Polysciences, Inc. Supplier No. 246331
Von Kossa Method for Calcium Kit demonstrates calcium phosphate and calcium carbonate salts. Von Kossa Method for Calcium Kit can reveal calcium deposits seen in metabolic diseases, Pagets disease, renal osteodystrophy, hyperparathyroidism, infarction (Gandy Gamna bodies), necrotic areas associated with TB, atheroma in blood vessels and Malakoplakia in the bladder (Michaelis-Gutman bodies). It is also used as a bone stain to indicate Osteomalacia.