Learn More
Selleck Chemical LLC Vonoprazan Fumarate
SDP
Catalog No. 5000001125
Encompass_Preferred
Vonoprazan Fumarate (TAK-438) is a novel P-CAB (potassium-competitive acid blocker) that reversibly inhibits H /K ATPase with IC50 of 19 nM (pH 6 5) controls gastric acid secretion Phase 3
Catalog No. 50-000-01125 Supplier Selleck Chemical LLC Supplier No. S80161G
Vonoprazan Fumarate (TAK-438) is a novel P-CAB (potassium-competitive acid blocker) that reversibly inhibits H /K ATPase with IC50 of 19 nM (pH 6 5) controls gastric acid secretion Phase 3