Learn More
Abcam Vonoprazan fumarate, 250MG
Catalog No. 502205862
Encompass_Preferred
MW 461.5 Da, Purity >=98%. Potassium-competitive acid blocker (P-CAB) that inhibits gastric H⁺/K⁺ATPase (IC₅₀ = 17 nM) but does not inhibit porcine Na+/K+ ATPase activity when used at a concentration of 10 μM.
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. 50-220-5862 Supplier Abcam Supplier No. AB286991250MG
MW 461.5 Da, Purity >=98%. Potassium-competitive acid blocker (P-CAB) that inhibits gastric H⁺/K⁺ATPase (IC₅₀ = 17 nM) but does not inhibit porcine Na+/K+ ATPase activity when used at a concentration of 10 μM.
The product is subject to the following: Abcam Restricted Use Statement