Learn More
Medchemexpress LLC Vonoprazan Fumarate (TAK-438) | 881681-01-2 | MFCD18633280 | C21H20FN3O6S | 1 ML
SDP
Catalog No. 5000372041
Encompass_Preferred
Vonoprazan Fumarate (TAK-438) is a potent and orally active potassium-competitive acid blocker (P-CAB) developed for research into acid-related diseases such as gastroesophageal reflux disease and peptic ulcer disease. It functions as a proton pump inhibitor (PPI) and exhibits antisecretory activity.
  • Potent and orally active potassium-competitive acid blocker (P-CAB)
  • Proton pump inhibitor (PPI)
  • Antisecretory activity
  • Inhibits H+,K+-ATPase activity with an IC50 of 19 nM at pH 6.5
Catalog No. 50-003-72041 Supplier Medchemexpress LLC Supplier No. HY1529510MM/1ML
Vonoprazan Fumarate (TAK-438) is a potent and orally active potassium-competitive acid blocker (P-CAB) developed for research into acid-related diseases such as gastroesophageal reflux disease and peptic ulcer disease. It functions as a proton pump inhibitor (PPI) and exhibits antisecretory activity.
  • Potent and orally active potassium-competitive acid blocker (P-CAB)
  • Proton pump inhibitor (PPI)
  • Antisecretory activity
  • Inhibits H+,K+-ATPase activity with an IC50 of 19 nM at pH 6.5