Learn More
Apexbio Technology LLC Vorapaxar 618385-01-6 10mM (in 1mL DMSO)
SDP
Catalog No. 5000061914
Encompass_Preferred
Vorapaxar (CAS 618385-01-6) is a selective inhibitor of protease-activated receptor 1 (PAR1) a G-protein coupled receptor activated by thrombin-mediated cleavage on platelet surfaces leading to platelet activation Derived from the natural product himbacine vorapaxar directly binds PAR1 inhibiting platelet aggregation induced by thrombin receptor-activating peptides (IC 25 nM) and thrombin itself (IC 47 nM) In phase III clinical trials in patients daily oral administration reduced the incidence of cardiovascular death or ischemic events highlighting its relevance as an antithrombotic research agent
Catalog No. 50-000-61914 Supplier Apexbio Technology LLC Supplier No. A88095.1
Vorapaxar (CAS 618385-01-6) is a selective inhibitor of protease-activated receptor 1 (PAR1) a G-protein coupled receptor activated by thrombin-mediated cleavage on platelet surfaces leading to platelet activation Derived from the natural product himbacine vorapaxar directly binds PAR1 inhibiting platelet aggregation induced by thrombin receptor-activating peptides (IC 25 nM) and thrombin itself (IC 47 nM) In phase III clinical trials in patients daily oral administration reduced the incidence of cardiovascular death or ischemic events highlighting its relevance as an antithrombotic research agent