Learn More
Apexbio Technology LLC Vorapaxar,5mg CAS# 618385-01-6
SDP
Supplier:  Apexbio Technology LLC A88095
Encompass_Preferred
Vorapaxar is a potent PAR1 inhibitor and antiplatelet that was completed in phase III clinical trial[1]. PAR1 is one of the 4 thrombin receptor types that belong to G-protein coupled receptors super family[1]. By binding to thrombin, the N-terminal at the arginine 41 and serine 42 bond of PAR1 on platelet surface can be enzymatically cleaved, resulting in the activation of platelet and subsequent hemostasis process[1]. Vorapaxar is an ethyl-carbamate molecule that derived from natural himbacine and has been determined to be a selective PAR 1 inhibitor through direct binding[2]. In human plasma enriched with platelets, Vorapaxar was able to inhibit the aggregation of platelet with an IC50 value of 47 nM[2]. In addition, vorapaxar is able to inhibitthe platelet aggregation that triggered by the addition of thrombin receptor activating peptide with an IC50 value of 25 nM[2]. Other sizes are also available. Please inqury us for quote.
Catalog No. 50-224-0144