Learn More
Medchemexpress LLC Vorapaxar sulfate | 705260-08-8 | MFCD16038877 | 99.8% | 590.66 g·mol⁻1 | C29H35FN2O8S | 10 MG
SDP
Catalog No. 5000212993
Encompass_Preferred
Vorapaxar sulfate is the sulfate salt form of vorapaxar, a selective protease-activated receptor-1 (PAR-1) antagonist used in research to study antiplatelet mechanisms. Supplied as a research reagent (commonly as a 10 mg solid vial), it is provided with documented chemical identifiers and high purity suitable for analytical and preclinical work.
  • Selective PAR-1 antagonist that inhibits platelet aggregation.
  • Sulfate salt form enhances stability and handling for assays.
  • High purity suitable for analytical and preclinical research.
  • Well characterized with CAS and database identifiers for traceability.
  • Molecular weight and formula available for formulation and dosing calculations.
Catalog No. 50-002-12993 Supplier Medchemexpress LLC Supplier No. HY10119A10MG
Vorapaxar sulfate is the sulfate salt form of vorapaxar, a selective protease-activated receptor-1 (PAR-1) antagonist used in research to study antiplatelet mechanisms. Supplied as a research reagent (commonly as a 10 mg solid vial), it is provided with documented chemical identifiers and high purity suitable for analytical and preclinical work.
  • Selective PAR-1 antagonist that inhibits platelet aggregation.
  • Sulfate salt form enhances stability and handling for assays.
  • High purity suitable for analytical and preclinical research.
  • Well characterized with CAS and database identifiers for traceability.
  • Molecular weight and formula available for formulation and dosing calculations.