Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Vorapaxar sulfate | 705260-08-8 | MFCD16038877 | 99.8% | 590.66 g/mol | C29H35FN2O8S | 5 MG
SDP

Catalog No. 5000213225
Encompass_Preferred

Vorapaxar sulfate is the sulfate salt of vorapaxar, a selective, orally active protease-activated receptor-1 (PAR-1) antagonist used in antiplatelet research. Supplied as a high-purity research chemical for biochemical, pharmacology, and assay development.

  • Selective PAR-1 antagonism for antiplatelet studies.
  • High purity, ≈99.8%.
  • Molecular formula C29H35FN2O8S; molecular weight 590.66 g/mol.
  • CAS number 705260-08-8 for unambiguous identification.
  • Available in small quantities, including 5 MG, for research use.
  • Intended for research and analytical applications only, not for human consumption.

Catalog No. 50-002-13225 Supplier Medchemexpress LLC Supplier No. HY10119A5MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Vorapaxar sulfate is the sulfate salt of vorapaxar, a selective, orally active protease-activated receptor-1 (PAR-1) antagonist used in antiplatelet research. Supplied as a high-purity research chemical for biochemical, pharmacology, and assay development.

  • Selective PAR-1 antagonism for antiplatelet studies.
  • High purity, ≈99.8%.
  • Molecular formula C29H35FN2O8S; molecular weight 590.66 g/mol.
  • CAS number 705260-08-8 for unambiguous identification.
  • Available in small quantities, including 5 MG, for research use.
  • Intended for research and analytical applications only, not for human consumption.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.