Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Vorapaxar sulfate (SCH 530348) | 705260-08-8 | MFCD16038877 | 99.8% | 590.66 g/mol | C29H35FN2O8S | 50 MG
SDP

Catalog No. 5000213386
Encompass_Preferred

Vorapaxar sulfate is the sulfate salt of vorapaxar, a selective protease-activated receptor 1 (PAR-1) antagonist with antiplatelet activity. It is supplied as a high-purity solid for research and analytical applications and is not intended for human or clinical use.

  • High purity: 99.81%.
  • Molecular weight: 590.66 g/mol.
  • Chemical formula: C29H35FN2O8S.
  • CAS number: 705260-08-8.
  • Available in multiple pack sizes, including 50 mg.
  • Solid form for straightforward handling and storage.

Catalog No. 50-002-13386 Supplier Medchemexpress LLC Supplier No. HY10119A50MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Vorapaxar sulfate is the sulfate salt of vorapaxar, a selective protease-activated receptor 1 (PAR-1) antagonist with antiplatelet activity. It is supplied as a high-purity solid for research and analytical applications and is not intended for human or clinical use.

  • High purity: 99.81%.
  • Molecular weight: 590.66 g/mol.
  • Chemical formula: C29H35FN2O8S.
  • CAS number: 705260-08-8.
  • Available in multiple pack sizes, including 50 mg.
  • Solid form for straightforward handling and storage.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.