Learn More
Cayman Chemical VorasIdenIb 10mg
SDP
Catalog No. 502941165
Encompass_Preferred
An inhibitor of mutant IDH (IC50s = 31.9 and 31.7 nM for IDH1R132H and IDH2R140Q, respectively); inhibits wild-type-IDH1R132H and wild-type-IDH2R140Q heterodimers (IC50s = 4 and 251 nM, respectively); decreases the production of 2-HG in TF-1 leukemia cells expressing IDH1R132H or IDH2R140Q (IC50s = 3.2 and 14 nM, respectively); reduces tumor growth in a TS603-IDH1R132H mouse xenograft model at 50 mg/kg
Catalog No. 50-294-1165 Supplier Cayman Chemical Supplier No. 2846210 MG
An inhibitor of mutant IDH (IC50s = 31.9 and 31.7 nM for IDH1R132H and IDH2R140Q, respectively); inhibits wild-type-IDH1R132H and wild-type-IDH2R140Q heterodimers (IC50s = 4 and 251 nM, respectively); decreases the production of 2-HG in TF-1 leukemia cells expressing IDH1R132H or IDH2R140Q (IC50s = 3.2 and 14 nM, respectively); reduces tumor growth in a TS603-IDH1R132H mouse xenograft model at 50 mg/kg