Learn More
Medchemexpress LLC HY-104042 10mg Medchemexpress, Vorasidenib CAS:1644545-52-7 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY10404210MG
Encompass_Preferred
Medchemexpress, HY-104042 10mg Vorasidenib CAS:1644545-52-7 Vorasidenib (AG-881) is an orally available, brain penetrant second-generation dual mutant isocitrate dehydrogenases 1 and 2 (mIDH1/2) inhibitor. Vorasidenib (AG-881) exhibits nanomolar inhibition of (D)-2-hydroxyglutarate (D-2-HG), and the IC50 ranges of 0.04~22 nM against IDH1 R132C, IDH1 R132G, IDH1 R132H and IDH1 R132S and 7~14 nM against IDH2 R140Q and 130 nM against IDH2 R172K[2]. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-202-8741