Learn More
Medchemexpress LLC Voreloxin hydrochloride | 175519-16-1 | 99.6% | 437.90 | 25 MG
SDP
Catalog No. 5000393058
Encompass_Preferred
Voreloxin Hydrochloride is a first-in-class topoisomerase II inhibitor that intercalates DNA and induces site-selective DNA double-strand breaks, G2 arrest, and apoptosis.
  • Induces G2 arrest partially through topoisomerase II in A549 lung cancer cell line.
  • Cytotoxic activity requires DNA intercalation.
  • Exhibits potent cytotoxic activity in AML cell lines, including MV4-11 and HL-60.
  • Shows additive or synergistic activity in acute leukemia cell lines when combined with cytarabine.
  • Causes accumulation of cells in the S and G2 phases of the cell cycle.
  • Induces bone marrow aplasia in vivo when combined with cytarabine.
Catalog No. 50-003-93058 Supplier Medchemexpress LLC Supplier No. 25MGHY16518
Voreloxin Hydrochloride is a first-in-class topoisomerase II inhibitor that intercalates DNA and induces site-selective DNA double-strand breaks, G2 arrest, and apoptosis.
  • Induces G2 arrest partially through topoisomerase II in A549 lung cancer cell line.
  • Cytotoxic activity requires DNA intercalation.
  • Exhibits potent cytotoxic activity in AML cell lines, including MV4-11 and HL-60.
  • Shows additive or synergistic activity in acute leukemia cell lines when combined with cytarabine.
  • Causes accumulation of cells in the S and G2 phases of the cell cycle.
  • Induces bone marrow aplasia in vivo when combined with cytarabine.