Learn More
TARGETMOL CHEMICALS INC Voriconazole 100MG
SDP
Catalog No. 502880755
Encompass_Preferred
Also available in 1 mL, 10 mg, 25 mg, 50 mg, 200 mg, 500 mg and bulk. Please contact Fisher for quotes. Voriconazole (UK-109496) is a triazole antifungal agent that specifically inhibits STEROL 14-ALPHA-DEMETHYLASE and CYTOCHROME P-450 [CYP3A]. Purity 99.44%
Catalog No. 50-288-0755 Supplier TARGETMOL CHEMICALS INC Supplier No. T0120100MG
Also available in 1 mL, 10 mg, 25 mg, 50 mg, 200 mg, 500 mg and bulk. Please contact Fisher for quotes. Voriconazole (UK-109496) is a triazole antifungal agent that specifically inhibits STEROL 14-ALPHA-DEMETHYLASE and CYTOCHROME P-450 [CYP3A]. Purity 99.44%