Learn More
Apexbio Technology LLC Voriconazole, 10mg. Cas: 137234-62-9 MFCD: MFCD00905717. CYP51 inhibitor
SDP
Supplier:  Apexbio Technology LLC A432010
Encompass_Preferred
Voriconazole is an inhibitor of 14-lanosterol demethylase with IC50 value of 53nM [1].Voriconazole is a triazole antifungal and a second-generation synthetic derivative of fluconazole. Other sizes are available. Please inqury us for quote.
Catalog No. 50-190-4920