Learn More
Selleck Chemical LLC Voriconazole S1442-10mM/1mL
SDP
Catalog No. 503111319
Encompass_Preferred
Voriconazole is a new triazole derivative similar to fluconazole and itraconazole that acts by inhibiting fungal cytochrome P-450-dependent 14-alpha-sterol demethylase-mediated synthesis of ergosterol
Catalog No. 50-311-1319 Supplier Selleck Chemical LLC Supplier No. S144210MM/1ML
Voriconazole is a new triazole derivative similar to fluconazole and itraconazole that acts by inhibiting fungal cytochrome P-450-dependent 14-alpha-sterol demethylase-mediated synthesis of ergosterol