Learn More
Medchemexpress LLC Voriconazole-d3 | 1217661-14-7 | 99.9% | C16H11D3F3N5O | 1 MG
SDP
Catalog No. 5000434654
Encompass_Preferred
Voriconazole-d3 is the deuterium labeled Voriconazole. Voriconazole (UK-109496) is a second-generation, broad-spectrum triazole antifungal agent that inhibits fungal ergosterol biosynthesis. It exerts its antifungal activity by inhibiting 14-α-lanosterol demethylation, a process mediated by fungal cytochrome P450 enzymes.
  • Can be used as a tracer.
  • Can be utilized as an internal standard for quantitative analysis through methods such as NMR, GC-MS, or LC-MS.
  • Stable heavy isotopes, including deuterium, have been incorporated into drug molecules as tracers for quantitation during drug development.
  • Deuteration can influence the pharmacokinetic and metabolic profiles of drugs.
Catalog No. 50-004-34654 Supplier Medchemexpress LLC Supplier No. HY76200S1MG
Voriconazole-d3 is the deuterium labeled Voriconazole. Voriconazole (UK-109496) is a second-generation, broad-spectrum triazole antifungal agent that inhibits fungal ergosterol biosynthesis. It exerts its antifungal activity by inhibiting 14-α-lanosterol demethylation, a process mediated by fungal cytochrome P450 enzymes.
  • Can be used as a tracer.
  • Can be utilized as an internal standard for quantitative analysis through methods such as NMR, GC-MS, or LC-MS.
  • Stable heavy isotopes, including deuterium, have been incorporated into drug molecules as tracers for quantitation during drug development.
  • Deuteration can influence the pharmacokinetic and metabolic profiles of drugs.