Learn More
Cayman Chemical VorIconazole-d3 N-oxIde 100ug
SDP
Catalog No. 502942962
Encompass_Preferred
An internal standard for the quantification of voriconazole N-oxide by GC- or LC-MS
Catalog No. 50-294-2962 Supplier Cayman Chemical Supplier No. 40332100 UG
An internal standard for the quantification of voriconazole N-oxide by GC- or LC-MS