Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Voriconazole impurity 1 | 137330-52-0 | ≥98.0% | C16H14F3N5O | 50 MG
SDP

Catalog No. 5000433898
Encompass_Preferred

Voriconazole impurity 1 is a chemical compound that serves as an impurity of Voriconazole. It has a molecular weight of 349.31 and a chemical formula of C16H14F3N5O. This solid substance has a purity of ≥98.0% and is intended for research use only.

  • Suitable for research applications
  • Purity of ≥98.0%
  • Solid appearance
  • Store at 4°C in sealed, moisture-free conditions
  • When in solvent, store at -80°C for 6 months or -20°C for 1 month

Catalog No. 50-004-33898 Supplier Medchemexpress LLC Supplier No. HY76200A50MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Voriconazole impurity 1 is a chemical compound that serves as an impurity of Voriconazole. It has a molecular weight of 349.31 and a chemical formula of C16H14F3N5O. This solid substance has a purity of ≥98.0% and is intended for research use only.

  • Suitable for research applications
  • Purity of ≥98.0%
  • Solid appearance
  • Store at 4°C in sealed, moisture-free conditions
  • When in solvent, store at -80°C for 6 months or -20°C for 1 month

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.