Learn More
Medchemexpress LLC Voriconazole (Standard) | 137234-62-9 | 99.9% | C16H14F3N5O | 25 MG
SDP
Catalog No. 5000358235
Encompass_Preferred
Voriconazole (Standard) is the analytical standard of Voriconazole, intended for research and analytical applications. It is a second-generation, broad-spectrum triazole antifungal agent that inhibits fungal ergosterol biosynthesis by inhibiting 14-α-lanosterol demethylation, a process mediated by fungal cytochrome P450 enzymes. This product serves as a reference standard for assay, commonly utilized in qualitative, quantitative, and methodological research experiments involving HPLC, GC, and MS.
  • Analytical standard grade
  • Inhibits fungal ergosterol biosynthesis
  • Broad-spectrum triazole antifungal agent
  • Used in HPLC, GC, and MS experiments
  • For research use only
  • Stable under recommended storage conditions
  • Not a hazardous substance or mixture
Catalog No. 50-003-58235 Supplier Medchemexpress LLC Supplier No. 25MGHY76200R
Voriconazole (Standard) is the analytical standard of Voriconazole, intended for research and analytical applications. It is a second-generation, broad-spectrum triazole antifungal agent that inhibits fungal ergosterol biosynthesis by inhibiting 14-α-lanosterol demethylation, a process mediated by fungal cytochrome P450 enzymes. This product serves as a reference standard for assay, commonly utilized in qualitative, quantitative, and methodological research experiments involving HPLC, GC, and MS.
  • Analytical standard grade
  • Inhibits fungal ergosterol biosynthesis
  • Broad-spectrum triazole antifungal agent
  • Used in HPLC, GC, and MS experiments
  • For research use only
  • Stable under recommended storage conditions
  • Not a hazardous substance or mixture