Learn More
Medchemexpress LLC Voriconazole (Standard) | 137234-62-9 | 99.90% | C16H14F3N5O | 5 MG
SDP
Catalog No. 5000433855
Encompass_Preferred
Voriconazole (Standard) is the analytical standard of Voriconazole, intended for research and analytical applications. Voriconazole (UK-109496) is a second-generation, broad-spectrum triazole antifungal agent that inhibits fungal ergosterol biosynthesis. Its antifungal activity is exerted through the inhibition of 14-α-lanosterol demethylation, a process mediated by fungal cytochrome P450 enzymes.
  • Used in qualitative, quantitative, and methodological research experiments in HPLC, GC, and MS.
  • Analytical standard for research and analytical applications.
  • Inhibits fungal ergosterol biosynthesis.
  • Exerts antifungal activity by inhibiting 14-α-lanosterol demethylation.
Catalog No. 50-004-33855 Supplier Medchemexpress LLC Supplier No. HY76200R5MG
Voriconazole (Standard) is the analytical standard of Voriconazole, intended for research and analytical applications. Voriconazole (UK-109496) is a second-generation, broad-spectrum triazole antifungal agent that inhibits fungal ergosterol biosynthesis. Its antifungal activity is exerted through the inhibition of 14-α-lanosterol demethylation, a process mediated by fungal cytochrome P450 enzymes.
  • Used in qualitative, quantitative, and methodological research experiments in HPLC, GC, and MS.
  • Analytical standard for research and analytical applications.
  • Inhibits fungal ergosterol biosynthesis.
  • Exerts antifungal activity by inhibiting 14-α-lanosterol demethylation.