Learn More
Selleck Chemical LLC Voriconazole 10mg 137234-62-9 UK-109496
SDP
Catalog No. 507319
Encompass_Preferred
Voriconazole (UK-109496) is a new triazole derivative similar to fluconazole and itraconazole that acts by inhibiting fungal cytochrome P-450-dependent, 14-alpha-sterol demethylase-mediated synthesis of ergosterol. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. 50-731-9 Supplier Selleck Chemical LLC Supplier No. S144210MG
Voriconazole (UK-109496) is a new triazole derivative similar to fluconazole and itraconazole that acts by inhibiting fungal cytochrome P-450-dependent, 14-alpha-sterol demethylase-mediated synthesis of ergosterol. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.